Order Entry
Sweden
ContactUsLinkComponent
Mouse VEGF R2, Mouse IgG2a Fc Tag, low endotoxin (VE2-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 762 (Accession P35918-1). 1 * 100 µG
Catalog # ACRBVE2-M5258100UG
Supplier:  ACRO Biosystems
Mouse VEGF R2, Mouse IgG2a Fc Tag, low endotoxin (VE2-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 762 (Accession P35918-1). 1 * 100 µG
Catalog # ACRBVE2-M5258100UG
Supplier:  ACRO Biosystems
Frequently Bought Together