Order Entry
Sweden
ContactUsLinkComponent
Human CD117, Fc Tag (CD7-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession P10721-2). 1 * 1 mg
Catalog # ACRBCD7-H5255-1MG
Leverantör:  ACRO Biosystems
Human CD117, Fc Tag (CD7-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession P10721-2). 1 * 1 mg
Catalog # ACRBCD7-H5255-1MG
Leverantör:  ACRO Biosystems
Frequently Bought Together