Order Entry
Sweden
ContactUsLinkComponent
Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (B75-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 1 94 (Accession AAH20568.1). 1 * 100 µG
Catalog # ACRBB75-H5258100UG
Leverantör:  ACRO Biosystems
Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (B75-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 1 94 (Accession AAH20568.1). 1 * 100 µG
Catalog # ACRBB75-H5258100UG
Leverantör:  ACRO Biosystems
Frequently Bought Together