Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (FC4-M82E8) is expressed from human 293 cells (HEK293). It contains AA Gl y 21 - Gln 203 (Accession A0A0B4J1G0-1). 1 * 25 µG
Catalog # ACRBFC4-M82E8-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (FC4-M82E8) is expressed from human 293 cells (HEK293). It contains AA Gl y 21 - Gln 203 (Accession A0A0B4J1G0-1). 1 * 25 µG
Catalog # ACRBFC4-M82E8-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together