0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 µG
ACRBTN5-M5259100UG
:
ACROBIOSYSTEMS
Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 µG
ACRBTN5-M5259100UG
:
ACROBIOSYSTEMS