Order Entry
Sweden
ContactUsLinkComponent
Mouse ICOS (C137S, C138S), Fc Tag (ICS-M5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accession Q9WVS0-1 (C137S, C138S)). 1 * 100 µG
Catalog # ACRBICS-M5259100UG
Supplier:  ACRO Biosystems
Mouse ICOS (C137S, C138S), Fc Tag (ICS-M5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accession Q9WVS0-1 (C137S, C138S)). 1 * 100 µG
Catalog # ACRBICS-M5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together