Order Entry
Sweden
ContactUsLinkComponent
Recombinat Protein,Human HABP1 C1QBP Protein, His Tag,Human HABP1, His Tag (C1P-H5122) is expressed from E.coli cells. It contains A A His 75 - Gln 282 (Accession AAH00435).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBC1P-H5122-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human HABP1 C1QBP Protein, His Tag,Human HABP1, His Tag (C1P-H5122) is expressed from E.coli cells. It contains A A His 75 - Gln 282 (Accession AAH00435).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBC1P-H5122-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together