Order Entry
Sweden
ContactUsLinkComponent
Recombinat Protein,Human Apolipoprotein A-II APOA2 Protein, His Tag,Human Apolipoprotein A-II, His Tag (APA-H51H6) is expressed from E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the C 1 * 1 mg
Catalog # ACRBAPA-H51H6-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Apolipoprotein A-II APOA2 Protein, His Tag,Human Apolipoprotein A-II, His Tag (APA-H51H6) is expressed from E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the C 1 * 1 mg
Catalog # ACRBAPA-H51H6-1MG
Supplier:  ACROBIOSYSTEMS