0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Human GM-CSF R alpha, Fc Tag (GRA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P155 09-1). 1 * 1 mg
ACRBGRA-H5255-1MG
:
ACROBIOSYSTEMS
Human GM-CSF R alpha, Fc Tag (GRA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P155 09-1). 1 * 1 mg
ACRBGRA-H5255-1MG
:
ACROBIOSYSTEMS