Order Entry
Sweden
ContactUsLinkComponent
HIV-1 HIV-1Clade C (16055) GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 ( Accession ABL67444.1). 1 * 1 mg
Catalog # ACRBGP5-V15224-1MG
Supplier:  ACROBIOSYSTEMS
HIV-1 HIV-1Clade C (16055) GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 ( Accession ABL67444.1). 1 * 1 mg
Catalog # ACRBGP5-V15224-1MG
Supplier:  ACROBIOSYSTEMS