Order Entry
Sweden
ContactUsLinkComponent
Human LRP-6 (20-630), Mouse IgG2a Fc Tag (LR6-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Ac cession O75581-1 ). 1 * 1 mg
Catalog # ACRBLR6-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Human LRP-6 (20-630), Mouse IgG2a Fc Tag (LR6-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Ac cession O75581-1 ). 1 * 1 mg
Catalog # ACRBLR6-H5253-1MG
Supplier:  ACROBIOSYSTEMS