Order Entry
Sweden
ContactUsLinkComponent
Human SIRP alpha, Fc Tag (HPLC-verified) (SIA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Ac cession NP001035111). 1 * 100 µG
Catalog # ACRBSIA-H5251100UG
Supplier:  ACROBIOSYSTEMS
Human SIRP alpha, Fc Tag (HPLC-verified) (SIA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Ac cession NP001035111). 1 * 100 µG
Catalog # ACRBSIA-H5251100UG
Supplier:  ACROBIOSYSTEMS