Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Human PD-1, Avitag,His Tag (recommended for biopanning) (PD1-H82E4) is expressed from human 293 cells (HEK293). It cont ains AA Leu 25 - Gln 167 (Accession Q15116-1). 1 * 200 µG
Catalog # ACRBPD1-H82E4200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human PD-1, Avitag,His Tag (recommended for biopanning) (PD1-H82E4) is expressed from human 293 cells (HEK293). It cont ains AA Leu 25 - Gln 167 (Accession Q15116-1). 1 * 200 µG
Catalog # ACRBPD1-H82E4200UG
Supplier:  ACROBIOSYSTEMS