Order Entry
Sweden
ContactUsLinkComponent
Cynomolgus SIRP alpha, His Tag (SIA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 370 (Accession NP 001271679.1). 1 * 100 µG
Catalog # ACRBSIA-C52H7100UG
Supplier:  ACROBIOSYSTEMS
Cynomolgus SIRP alpha, His Tag (SIA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 370 (Accession NP 001271679.1). 1 * 100 µG
Catalog # ACRBSIA-C52H7100UG
Supplier:  ACROBIOSYSTEMS