Order Entry
Sweden
ContactUsLinkComponent
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 100 µG
Catalog # ACRBCEB-Y31-100UG
Leverantör:  ACROBIOSYSTEMS
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 100 µG
Catalog # ACRBCEB-Y31-100UG
Leverantör:  ACROBIOSYSTEMS