Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P40933-1). 1 * 200 µG
Catalog # ACRBIL5-H82F3200UG
Leverantör:  ACROBIOSYSTEMS
Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P40933-1). 1 * 200 µG
Catalog # ACRBIL5-H82F3200UG
Leverantör:  ACROBIOSYSTEMS