Order Entry
Sweden
ContactUsLinkComponent
Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (SIA-M5252) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession BAA13520.1 (G365D)). 1 * 100 µG
Catalog # ACRBSIA-M5252100UG
Supplier:  ACRO Biosystems
Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (SIA-M5252) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession BAA13520.1 (G365D)). 1 * 100 µG
Catalog # ACRBSIA-M5252100UG
Supplier:  ACRO Biosystems
Frequently Bought Together