Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (SIA-M5252) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession BAA13520.1 (G365D)). 1 * 1 mg
Catalog # ACRBSIA-M5252-1MG
Leverantör: ACRO Biosystems
Frequently Bought Together