Order Entry
Sweden
ContactUsLinkComponent
Human CD19 (20-291), Mouse IgG2a Fc Tag (CD9-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Acc ession P15391-1). 1 * 100 µG
Catalog # ACRBCD9-H5258100UG
Supplier:  ACRO Biosystems
Human CD19 (20-291), Mouse IgG2a Fc Tag (CD9-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Acc ession P15391-1). 1 * 100 µG
Catalog # ACRBCD9-H5258100UG
Supplier:  ACRO Biosystems
Frequently Bought Together