Order Entry
Sweden
ContactUsLinkComponent
Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (OXL-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession F7FL80-1). 1 * 100 µG
  ACRBOXL-R5259100UG
Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (OXL-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession F7FL80-1). 1 * 100 µG
  ACRBOXL-R5259100UG
Frequently Bought Together