Order Entry
Sweden
ContactUsLinkComponent
Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (BCA-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession Q02223-1). 1 * 100 µG
Catalog # ACRBBCA-H5253100UG
Supplier:  ACRO Biosystems
Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (BCA-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession Q02223-1). 1 * 100 µG
Catalog # ACRBBCA-H5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together