Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Human Siglec-15, Fc,Avitag (SG5-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Acc ession Q6ZMC9-1). 1 * 200 µG
  ACRBSG5-H82F5200UG
Biotinylated Human Siglec-15, Fc,Avitag (SG5-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Acc ession Q6ZMC9-1). 1 * 200 µG
  ACRBSG5-H82F5200UG
Frequently Bought Together