Order Entry
Sweden
ContactUsLinkComponent
Cynomolgus Rhesus macaque FGL1, Mouse IgG2a Fc Tag (FG1-C5250) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - I le 312 (Accession G7N0K6-1). 1 * 100 µG
Catalog # ACRBFG1-C5250100UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque FGL1, Mouse IgG2a Fc Tag (FG1-C5250) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - I le 312 (Accession G7N0K6-1). 1 * 100 µG
Catalog # ACRBFG1-C5250100UG
Supplier:  ACRO Biosystems
Frequently Bought Together