Antibody,Anti-Trastuzumab Antibodies (recommended for PKPD),Anti-Trastuzumab Antibodies (recommended for PKPD) (TRB-Y5b) are express ed from human 293 cells (HEK293). ,Immobilized Trastuzumab at 1 ugmL (100 uLwell) can bind Anti-Trastuzumab Antibodie 1 * 100 µG
Catalog # ACRBTRB-Y5B-100UG
Supplier: ACRO Biosystems
Frequently Bought Together