0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
pH electrode, plastic/gel, type 226, without temperature sensor, BNC-plug 1 * 1 items
LOVI721226
:
Lovibond
pH electrode, plastic/gel, type 226, without temperature sensor, BNC-plug 1 * 1 items
LOVI721226
:
Lovibond