Order Entry
Sweden
ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 75x75cm, DIN EN ISO 11607 1 * 50 items
Catalog # VEPA3FDOP383208
Leverantör:  VP Medical Packaging
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 75x75cm, DIN EN ISO 11607 1 * 50 items
Catalog # VEPA3FDOP383208
Leverantör:  VP Medical Packaging
Frequently Bought Together