0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LC BULK MEDIA 1 60A 40-63 UM DAVISIL 1 * 1 kg
Catalog # HISM5183335
Supplier:
HICHROM
Expand image
LC BULK MEDIA 1 60A 40-63 UM DAVISIL 1 * 1 kg
Catalog # HISM5183335
Supplier:
HICHROM