0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REPROSIL CHIRAL-BETA-PM 5 µM 250 X 4.6 1 * 1 items
Catalog # MAISR15.BPM.S2546
Supplier:
DR MAISCH
Expand image
REPROSIL CHIRAL-BETA-PM 5 µM 250 X 4.6 1 * 1 items
Catalog # MAISR15.BPM.S2546
Supplier:
DR MAISCH