0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACHIRAL ANALYTICAL COLUMN DAICEL DCpak® PTZ 150 x 3MM 3 micron 1 * 1 items
Catalog # DAICAC584
Supplier:
CHIRAL TECHNOLOGIES
Expand image
ACHIRAL ANALYTICAL COLUMN DAICEL DCpak® PTZ 150 x 3MM 3 micron 1 * 1 items
Catalog # DAICAC584
Supplier:
CHIRAL TECHNOLOGIES