Order Entry
Sweden
ContactUsLinkComponent
A 10X CONCENTRATED RED BLOOD CELL LYSIS BUFFER FOR THE ISOLATION OF WHITE BLOOD CELLS FREE FROM INTERFERENCE OF RBC. THE OPTIMIZED CONCENTRATION OF AMMONIUM CHLORIDE AND ENHANCES EFFECTIVELY LYSES RED BLOOD 1 * 100 mL
Catalog # GENO786-1701
Supplier:  G-Biosciences
A 10X CONCENTRATED RED BLOOD CELL LYSIS BUFFER FOR THE ISOLATION OF WHITE BLOOD CELLS FREE FROM INTERFERENCE OF RBC. THE OPTIMIZED CONCENTRATION OF AMMONIUM CHLORIDE AND ENHANCES EFFECTIVELY LYSES RED BLOOD 1 * 100 mL
Catalog # GENO786-1701
Supplier:  G-Biosciences
Frequently Bought Together