Order Entry
Sweden
ContactUsLinkComponent
Cell Cryopreservation Medium with 10% DMSO is a balanced, protein free, ready-to-use cryopreservation solution with 10% USP DMSO (Cr yopres&trade ). 1 * 50 mL
Catalog # 092780248.
Supplier:  MP Biomedicals
Cell Cryopreservation Medium with 10% DMSO is a balanced, protein free, ready-to-use cryopreservation solution with 10% USP DMSO (Cr yopres&trade ). 1 * 50 mL
Catalog # 092780248.
Supplier:  MP Biomedicals
Frequently Bought Together