Order Entry
Sweden
ContactUsLinkComponent
Mouse IgG monoclonal antibody for FH detection. Tested with WB, IHC-P in Human Mouse Rat Monkey. 1 * 100 µG
Catalog # BSBTM02097
Supplier:  Boster Bio
Mouse IgG monoclonal antibody for FH detection. Tested with WB, IHC-P in Human Mouse Rat Monkey. 1 * 100 µG
Catalog # BSBTM02097
Supplier:  Boster Bio