Order Entry
Sweden
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for SOX9 detection. Tested with WB, IHC-P in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA00177-2
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for SOX9 detection. Tested with WB, IHC-P in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA00177-2
Supplier:  Boster Bio