Order Entry
Sweden
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Troponin I fast skeletal muscle/TNNI2 detection. Tested with WB, IHC-P, Direct ELISA in Human Mou se Rat. 1 * 100 µG
Catalog # BSBTA07355-2
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Troponin I fast skeletal muscle/TNNI2 detection. Tested with WB, IHC-P, Direct ELISA in Human Mou se Rat. 1 * 100 µG
Catalog # BSBTA07355-2
Supplier:  Boster Bio