Order Entry
Sweden
ContactUsLinkComponent
Detect Human Plasminogen/PLG with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: serum, pla sma and cell culture supernates. 1 * 1 KIT
Catalog # BSBTFEK0905
Supplier:  Boster Bio
Detect Human Plasminogen/PLG with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: serum, pla sma and cell culture supernates. 1 * 1 KIT
Catalog # BSBTFEK0905
Supplier:  Boster Bio