Order Entry
Sweden
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Cytochrome P450 2E1/CYP2E1 detection. Tested with WB, IHC-P, FCM, Direct ELISA in Human Mouse Rat . 1 * 100 µG
Catalog # BSBTA00672
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Cytochrome P450 2E1/CYP2E1 detection. Tested with WB, IHC-P, FCM, Direct ELISA in Human Mouse Rat . 1 * 100 µG
Catalog # BSBTA00672
Supplier:  Boster Bio