0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Volumetric pipette, 15ml, Class AS, DIN EN ISO 648 1 * 12 items
Catalog # WITG3.535.150.BL
Supplier:
witeg Labortechnik
Expand image
Volumetric pipette, 15ml, Class AS, DIN EN ISO 648 1 * 12 items
Catalog # WITG3.535.150.BL
Supplier:
witeg Labortechnik