0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HYPERSIL CHIRAL-AT 10UM 150X4.6MM COLUMN 1 * 1 items
Catalog # 555-0654
Supplier:
Thermo Fisher Scientific
Expand image
HYPERSIL CHIRAL-AT 10UM 150X4.6MM COLUMN 1 * 1 items
Catalog # 555-0654
Supplier:
Thermo Fisher Scientific