Order Entry
Sweden
ContactUsLinkComponent
Multiple Distributors for Bottles EX, Black screw cap made of conductive PPS-EX for bottle thread gl45, distributor body made of con ductive PTFE-EX with GL-threaded necks. Tubes can be inserted through the necks. Connection of tubes or tubing by mea 1 * 1 items
Catalog # BOHLD866-08
Leverantör:  Bohlender
undefined
Multiple Distributors for Bottles EX, Black screw cap made of conductive PPS-EX for bottle thread gl45, distributor body made of con ductive PTFE-EX with GL-threaded necks. Tubes can be inserted through the necks. Connection of tubes or tubing by mea 1 * 1 items
Catalog # BOHLD866-08
Leverantör:  Bohlender
Frequently Bought Together