Order Entry
Sweden
ContactUsLinkComponent
Wisp1 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat Wisp1 in cell culture supern ates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5466
Supplier:  Abnova
Wisp1 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat Wisp1 in cell culture supern ates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5466
Supplier:  Abnova