Order Entry
Sweden
ContactUsLinkComponent
SMOC2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human SMOC2 in cell culture su pernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  ABNOKA5460
 :  Abnova
SMOC2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human SMOC2 in cell culture su pernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  ABNOKA5460
 :  Abnova