Order Entry
Sweden
ContactUsLinkComponent
SENSOR CHIPS 5 SPR BIACORE SERIES S compatible.Coating: 200 nm streptavidin derivatized carboxymethyldextran hydrogel, medium densit y 1 * 5 items
Catalog # MSPPSCBS-SAD200M-5
SENSOR CHIPS 5 SPR BIACORE SERIES S compatible.Coating: 200 nm streptavidin derivatized carboxymethyldextran hydrogel, medium densit y 1 * 5 items
Catalog # MSPPSCBS-SAD200M-5
Frequently Bought Together