0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
pH/temp. electrode, type 231, plastic/gel/temp. NTC30kOhm, BNC-plug for SD 300 pH (SET 1) 1 * 1 items
LOVI721231
:
Lovibond
pH/temp. electrode, type 231, plastic/gel/temp. NTC30kOhm, BNC-plug for SD 300 pH (SET 1) 1 * 1 items
LOVI721231
:
Lovibond