Order Entry
Sweden
Orders LinkContactUsLinkComponent
SRK® Surfaces and Equipment Microbial Testing Devices

SRK® Surfaces and Equipment Microbial Testing Devices

SRK® - 1 ml SRK® solution in a sponge contained in a tube + 1 regular FLOQSwabs® + a separate tube filled; 50×6, individually wrapped; 139 mm choked tube/12×80 mm skirted conic bottom, 1 ml/1 ml, Green/green
Catalog # COPA9U012S
Supplier:   Copan
SRK® Surfaces and Equipment Microbial Testing Devices
Catalog # COPA9U012S
Supplier:  Copan
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® Surfaces and Equipment Microbial Testing Devices

SRK® - 1 ml SRK® solution in a sponge contained in a tube + 1 regular FLOQSwabs® + a separate tube filled; 50×6, individually wrapped; 139 mm choked tube/12×80 mm skirted conic bottom, 1 ml/1 ml, Green/green

Price
7 825,00 kr
Log in for availability
1 * 300 Items
Product Details & Documents

Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.

  • Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula

    Compliance: SRK® kits are designed compliant with ISO 18593:2018, ISO 18593:2004, ISO 17604:2003, and EU directive 2001/471/E.C

    Media choice for enhanced preservation: Choose between an isotonic solution or a wide range of selective and enrichment broths: Half Fraiser, RVS, BPW 1%, TSB, UNS, and Letheen broth

    Sterile sampling frames: the definition of a set sampling area is essential for microbial bioburden validations

    Sampling frames will make your surface sampling match that standard

This product line groups a variety of product codes, each optimised to address specific needs.

Specifications
  • Description:
    SRK® - 1 ml SRK® solution in a sponge contained in a tube + 1 regular FLOQSwabs® + a separate tube filled; 50×6, individually wrapped; 139 mm choked tube/12×80 mm skirted conic bottom, 1 ml/1 ml
  • Seq:
    12
  • Cap colour:
    Green/green
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.