0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Wachs Kegelnadel 2,5 g, standard, mit ID-Nummer, ASTM D 1321, DIN 51 579 1 * 1 ST
PAAR106843
:
Anton Paar
Wachs Kegelnadel 2,5 g, standard, mit ID-Nummer, ASTM D 1321, DIN 51 579 1 * 1 ST
PAAR106843
:
Anton Paar