Order Entry
Sweden
ContactUsLinkComponent
SDC3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human syndecan-3 in cell cultur e supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5185
Supplier:  Abnova
SDC3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human syndecan-3 in cell cultur e supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5185
Supplier:  Abnova