Order Entry
Sweden
ContactUsLinkComponent
Glove Box Glove Jugitec Pharma, Typ 17BL04 250F Material: EPDM Size: L (9-10), 0.4 mm / 16 mil thickness ambidextrous Gauntlet diameter: 250mm Type: 7BL04 250F Customs tariff no. 40151900 Marking: Jugitec® Pharma With FDA Certificate 1 * 1 PAIR
Catalog # BSNC17BL04250F
Supplier:  Basan
Glove Box Glove Jugitec Pharma, Typ 17BL04 250F Material: EPDM Size: L (9-10), 0.4 mm / 16 mil thickness ambidextrous Gauntlet diameter: 250mm Type: 7BL04 250F Customs tariff no. 40151900 Marking: Jugitec® Pharma With FDA Certificate 1 * 1 PAIR
Catalog # BSNC17BL04250F
Supplier:  Basan