Order Entry
Sweden
ContactUsLinkComponent
Stericlin® sterilisation reel, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, width 42cm x le ngth 100m, DIN EN ISO 11607 1 * 1 Roll
Catalog # 129-1295
Stericlin® sterilisation reel, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, width 42cm x le ngth 100m, DIN EN ISO 11607 1 * 1 Roll
Catalog # 129-1295