Order Entry
Sweden
ContactUsLinkComponent
Stericlin® container filter, made of paper, fits Aesculap containers, square with sloped edges, with STEAM indicator, size 230x230mm 1 * 500 items
Catalog # VEPA3FCZB470102
Stericlin® container filter, made of paper, fits Aesculap containers, square with sloped edges, with STEAM indicator, size 230x230mm 1 * 500 items
Catalog # VEPA3FCZB470102
Frequently Bought Together