Order Entry
Sweden
ContactUsLinkComponent
Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display 1 * 1 items
Catalog # 390-1259
Supplier:  Binder
Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display 1 * 1 items
Catalog # 390-1259
Supplier:  Binder
Frequently Bought Together